accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1415334-59-6

IUPAC Name: (10-{5-phenyl-[1,1'-biphenyl]-3-yl}anthracen-9-yl)boronic acid
Synonyms: 10-{5-phenyl-[1,1'-biphenyl]-3-yl}anthracen-9-ylboronic acid
Molecular Weight (g/mol): 450.34
Molecular Formula: C32H23BO2
InChi Key: NQOSAYPSOKFYBW-UHFFFAOYSA-N
SMILES: OB(O)C1=C2C=CC=CC2=C(C2=CC(=CC(=C2)C2=CC=CC=C2)C2=CC=CC=C2)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results