accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOH3CFFHO

CAS RN 141546-97-6

IUPAC Name: ethyl 3-ethoxy-2,2-difluoro-3-hydroxypropanoate
Synonyms: ethyl 3-ethoxy-2,2-difluoro-3-hydroxypropanoate
Molecular Weight (g/mol): 198.17
Molecular Formula: C7H12F2O4
InChi Key: DCDGEVPPIDIMON-UHFFFAOYNA-N
SMILES: CCOC(O)C(F)(F)C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results