accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNNClCl

CAS RN 14161-11-6

IUPAC Name: 3,4,5-trichloropyridazine
Synonyms: 3,4,5-trichloropyridazine
Molecular Weight (g/mol): 183.42
Molecular Formula: C4HCl3N2
InChi Key: GBAOOJAWDCNOGO-UHFFFAOYSA-N
SMILES: ClC1=CN=NC(Cl)=C1Cl
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)