accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOOOOOOOOH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OSmSm

CAS RN 14175-03-2

IUPAC Name: tris(oxalic acid) decahydrate disamarium
Synonyms: tris(oxalic acid) decahydrate disamarium
Molecular Weight (g/mol): 750.97
Molecular Formula: C6H26O22Sm2
InChi Key: IKPDQYODFRQYAD-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.O.O.O.O.[Sm].[Sm].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results