accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14177-46-9

IUPAC Name: manganese(2+) dioxotungstenbis(olate)
Synonyms: manganese(2+) tungstate
Molecular Weight (g/mol): 302.77
Molecular Formula: MnO4W
InChi Key: CRLHSBRULQUYOK-UHFFFAOYSA-N
SMILES: [Mn++].[O-][W]([O-])(=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results