accessibility menu, dialog, popup

UserName

Viewing profile as user:
NiOWOOO

CAS RN 14177-51-6

IUPAC Name: nickel(2+) dioxotungstenbis(olate)
Synonyms: nickel(2+) tungstate
Molecular Weight (g/mol): 306.53
Molecular Formula: NiO4W
InChi Key: QLTKZXWDJGMCAR-UHFFFAOYSA-N
SMILES: [Ni++].[O-][W]([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results