accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1418022-42-0

IUPAC Name: N-(35-amino-3,6,9,12,15,18,21,24,27,30,33-undecaoxapentatriacontan-1-yl)-5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanamide
Synonyms: N-(35-amino-3,6,9,12,15,18,21,24,27,30,33-undecaoxapentatriacontan-1-yl)-5-{2-oxo-hexahydrothieno[3,4-d]imidazol-4-yl}pentanamide
Molecular Weight (g/mol): 770.98
Molecular Formula: C34H66N4O13S
InChi Key: SHPHAOSBFNWGAC-UHFFFAOYNA-N
SMILES: NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(=O)CCCCC1SCC2NC(=O)NC12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results