CAS RN 14188-94-4
CAS RN 14188-94-4
IUPAC Name:
3-acetylpyridin-1-ium-1-olate
Synonyms:
3-acetylpyridin-1-ium-1-olate
Molecular Weight (g/mol):
137.14
Molecular Formula:
C7H7NO2
InChi Key:
AZFMFGIMDGBJRR-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC=C[N+]([O-])=C1
1
–
1
of
1
results
1
3-Acetylpyridine N-Oxide 98.0+%, TCI America™
CAS: 14188-94-4 Molecular Formula: C7H7NO2 Molecular Weight (g/mol): 137.138 MDL Number: MFCD00955798 InChI Key: AZFMFGIMDGBJRR-UHFFFAOYSA-N PubChem CID: 123338 IUPAC Name: 1-(1-oxidopyridin-1-ium-3-yl)ethanone SMILES: CC(=O)C1=C[N+](=CC=C1)[O-]