accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 142-03-0

IUPAC Name: (acetyloxy)(hydroxy)alumanyl acetate
Synonyms: aluminium diacetate|aluminum diacetate
Molecular Weight (g/mol): 162.08
Molecular Formula: C4H7AlO5
InChi Key: HQQUTGFAWJNQIP-UHFFFAOYSA-K
SMILES: CC(=O)O[Al](O)OC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)