accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 142-04-1

IUPAC Name: hydrogen aniline chloride
Synonyms: hydrogen aniline chloride
Molecular Weight (g/mol): 129.59
Molecular Formula: C6H8ClN
InChi Key: MMCPOSDMTGQNKG-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=CC=CC=C1
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (6)
  • (2)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)