accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 142-31-4

IUPAC Name: sodium octyl sulfate
Synonyms: sodium octyl sulfate
Molecular Weight (g/mol): 232.27
Molecular Formula: C8H17NaO4S
InChi Key: WFRKJMRGXGWHBM-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCCCOS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results