accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 142-72-3

IUPAC Name: magnesium(2+) diacetate
Synonyms: magnesium acetate magnesium di(acetate) acetic acid, magnesium salt (2:1) acetic acid magnesium salt mg acetate mg(II) acetate magnesium diacetate
Molecular Weight (g/mol): 142.39
Molecular Formula: C4H6MgO4
InChi Key: UEGPKNKPLBYCNK-UHFFFAOYSA-L
SMILES: [Mg++].CC([O-])=O.CC([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results