accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 142-88-1

IUPAC Name: hexanedioic acid; piperazine
Synonyms: adipic acid; piperazine
Molecular Weight (g/mol): 232.28
Molecular Formula: C10H20N2O4
InChi Key: BVEGEKOBSPXUJS-UHFFFAOYSA-N
SMILES: C1CNCCN1.OC(=O)CCCCC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results