accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3CH2

CAS RN 142-90-5

IUPAC Name: dodecyl 2-methylprop-2-enoate
Synonyms: dodecyl methacrylate
Molecular Weight (g/mol): 254.41
Molecular Formula: C16H30O2
InChi Key: GMSCBRSQMRDRCD-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCOC(=O)C(C)=C
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (6)
  • (2)
  • (3)