accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3O

CAS RN 142-92-7

IUPAC Name: hexyl acetate
Synonyms: hexyl acetate N-hexyl acetate
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: AOGQPLXWSUTHQB-UHFFFAOYSA-N
SMILES: CCCCCCOC(C)=O
Boiling Point 
  • (2)
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (4)
  • (2)
  • (7)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (5)