accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14205-39-1

IUPAC Name: methyl (2E)-3-aminobut-2-enoate
Synonyms: methyl (2E)-3-aminobut-2-enoate
Molecular Weight (g/mol): 115.13
Molecular Formula: C5H9NO2
InChi Key: XKORCTIIRYKLLG-ONEGZZNKSA-N
SMILES: COC(=O)\C=C(/C)N
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (3)
  • (5)