accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14205-47-1

IUPAC Name: 4-{[(2E)-3-aminobut-2-enoyl]oxy}butyl (2E)-3-aminobut-2-enoate
Synonyms: 4-{[(2E)-3-aminobut-2-enoyl]oxy}butyl (2E)-3-aminobut-2-enoate
Molecular Weight (g/mol): 256.3
Molecular Formula: C12H20N2O4
InChi Key: DFUXQARZEWMFLQ-FIFLTTCUSA-N
SMILES: C\C(N)=C/C(=O)OCCCCOC(=O)\C=C(/C)N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results