accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14206-58-7

IUPAC Name: 4-(dimethylamino)-1,5,6,10,11,12a-hexahydroxy-6-methyl-3,12-dioxo-3,4,4a,5,5a,6,12,12a-octahydrotetracene-2-carboxamide
Synonyms: 4-(dimethylamino)-1,5,6,10,11,12a-hexahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide
Molecular Weight (g/mol): 460.44
Molecular Formula: C22H24N2O9
InChi Key: OWFJMIVZYSDULZ-UHFFFAOYNA-N
SMILES: CN(C)C1C2C(O)C3C(=C(O)C4=C(C=CC=C4O)C3(C)O)C(=O)C2(O)C(O)=C(C(N)=O)C1=O
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)