accessibility menu, dialog, popup

UserName

Viewing profile as user:
NS

CAS RN 1421261-24-6

IUPAC Name: 4-{[1,1'-biphenyl]-4-yl}-2-cyclopropyl-1,3-thiazole
Synonyms: 4-{[1,1'-biphenyl]-4-yl}-2-cyclopropyl-1,3-thiazole
Molecular Weight (g/mol): 277.39
Molecular Formula: C18H15NS
InChi Key: ZGFVNJMOZOGCDK-UHFFFAOYSA-N
SMILES: C1CC1C1=NC(=CS1)C1=CC=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results