accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNClClBr

CAS RN 1421261-47-3

IUPAC Name: 3-(4-bromophenyl)-4-(3,4-dichlorophenyl)-5-methyl-4H-1,2,4-triazole
Synonyms: 3-(4-bromophenyl)-4-(3,4-dichlorophenyl)-5-methyl-1,2,4-triazole
Molecular Weight (g/mol): 383.07
Molecular Formula: C15H10BrCl2N3
InChi Key: YHMKSEQFQPFOLL-UHFFFAOYSA-N
SMILES: CC1=NN=C(N1C1=CC(Cl)=C(Cl)C=C1)C1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results