accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNOON

CAS RN 1421262-17-0

IUPAC Name: 5-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)pyridine-2-carbonitrile
Synonyms: 5-(2,5-dioxopyrrol-1-yl)pyridine-2-carbonitrile
Molecular Weight (g/mol): 199.17
Molecular Formula: C10H5N3O2
InChi Key: ORPMTHPTAABMBU-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)N1C1=CN=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results