accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOHNBrClCl

CAS RN 1421262-63-6

IUPAC Name: N-(2-bromo-4,6-dichlorophenyl)benzenesulfonamide
Synonyms: N-(2-bromo-4,6-dichlorophenyl)benzenesulfonamide
Molecular Weight (g/mol): 381.07
Molecular Formula: C12H8BrCl2NO2S
InChi Key: UYPWKTWHHVRHIG-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(NS(=O)(=O)C2=CC=CC=C2)C(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results