CAS RN 1421262-65-8
IUPAC Name:
4-(3-ethyl-1,2,4-oxadiazol-5-yl)benzonitrile
Synonyms:
4-(3-ethyl-1,2,4-oxadiazol-5-yl)benzonitrile
Molecular Weight (g/mol):
199.21
Molecular Formula:
C11H9N3O
InChi Key:
OLEJLLUUIDNJDM-UHFFFAOYSA-N
SMILES:
CCC1=NOC(=N1)C1=CC=C(C=C1)C#N
1
–
1
of
1
results
1
4-(3-Ethyl-1,2,4-oxadiazol-5-yl)benzonitrile, 97%, Thermo Scientific™
CAS: 1421262-65-8 Molecular Formula: C11H9N3O Molecular Weight (g/mol): 199.213 MDL Number: MFCD22682905 InChI Key: OLEJLLUUIDNJDM-UHFFFAOYSA-N Synonym: 4-3-ethyl-1,2,4-oxadiazol-5-yl benzonitrile PubChem CID: 73996265 IUPAC Name: 4-(3-ethyl-1,2,4-oxadiazol-5-yl)benzonitrile SMILES: CCC1=NOC(=N1)C2=CC=C(C=C2)C#N