CAS RN 1421262-75-0
IUPAC Name:
5-(4-{[1,1'-biphenyl]-4-yl}-1,3-thiazol-2-yl)-2-methylaniline
Synonyms:
5-(4-{[1,1'-biphenyl]-4-yl}-1,3-thiazol-2-yl)-2-methylaniline
Molecular Weight (g/mol):
342.46
Molecular Formula:
C22H18N2S
InChi Key:
SLRVZGOVKVVJAX-UHFFFAOYSA-N
SMILES:
CC1=C(N)C=C(C=C1)C1=NC(=CS1)C1=CC=C(C=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
2-(3-Amino-4-methylphenyl)-4-(4-biphenylyl)thiazole, 97%, Thermo Scientific™
CAS: 1421262-75-0 Molecular Formula: C22H18N2S Molecular Weight (g/mol): 342.46 MDL Number: MFCD22683000 InChI Key: SLRVZGOVKVVJAX-UHFFFAOYSA-N Synonym: 5-4-1,1'-biphenyl-4-yl-1,3-thiazol-2-yl-2-methylaniline,2-3-amino-4-methylphenyl-4-4-biphenylyl thiazole PubChem CID: 97030358 IUPAC Name: 2-methyl-5-[4-(4-phenylphenyl)-1,3-thiazol-2-yl]aniline SMILES: CC1=C(C=C(C=C1)C2=NC(=CS2)C3=CC=C(C=C3)C4=CC=CC=C4)N