accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1421262-98-7

H3CNS

CAS RN 1421262-98-7

IUPAC Name: 2-(4-methylphenyl)-4-(5,6,7,8-tetrahydronaphthalen-2-yl)-1,3-thiazole
Synonyms: 2-(4-methylphenyl)-4-(5,6,7,8-tetrahydronaphthalen-2-yl)-1,3-thiazole
Molecular Weight (g/mol): 305.44
Molecular Formula: C20H19NS
InChi Key: SZRKBHGWZOAMML-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=NC(=CS1)C1=CC2=C(CCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results