accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1421263-21-9

IUPAC Name: N-benzyl-N-tert-butyl-2,6-difluorobenzamide
Synonyms: N-benzyl-N-tert-butyl-2,6-difluorobenzamide
Molecular Weight (g/mol): 303.35
Molecular Formula: C18H19F2NO
InChi Key: IOQMLEAYZOPPSC-UHFFFAOYSA-N
SMILES: CC(C)(C)N(CC1=CC=CC=C1)C(=O)C1=C(F)C=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results