CAS RN 1421263-26-4
IUPAC Name:
4-{2-[(pyridin-3-yl)methyl]-1,3-thiazol-4-yl}pyridine
Synonyms:
4-[2-(pyridin-3-ylmethyl)-1,3-thiazol-4-yl]pyridine
Molecular Weight (g/mol):
253.32
Molecular Formula:
C14H11N3S
InChi Key:
KUCSSSQZPFTBHS-UHFFFAOYSA-N
SMILES:
C(C1=NC(=CS1)C1=CC=NC=C1)C1=CN=CC=C1
1
–
1
of
1
results
1
4-(4-Pyridyl)-2-(3-pyridylmethyl)thiazole, 97%, Thermo Scientific™
CAS: 1421263-26-4 Molecular Formula: C14H11N3S Molecular Weight (g/mol): 253.323 MDL Number: MFCD22682928 InChI Key: KUCSSSQZPFTBHS-UHFFFAOYSA-N Synonym: 4-4-pyridyl-2-3-pyridylmethyl thiazole,4-2-pyridin-3-ylmethyl-1,3-thiazol-4-yl pyridine,2-pyridin-3-ylmethyl-4-pyridin-4-yl thiazole PubChem CID: 71741367 IUPAC Name: 4-pyridin-4-yl-2-(pyridin-3-ylmethyl)-1,3-thiazole SMILES: C1=CC(=CN=C1)CC2=NC(=CS2)C3=CC=NC=C3