accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1421263-53-7

IUPAC Name: 3-(4-bromophenyl)-4-(4-ethylphenyl)-5-methyl-4H-1,2,4-triazole
Synonyms: 3-(4-bromophenyl)-4-(4-ethylphenyl)-5-methyl-1,2,4-triazole
Molecular Weight (g/mol): 342.24
Molecular Formula: C17H16BrN3
InChi Key: FSFAVMUCBYVKSQ-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=C1)N1C(C)=NN=C1C1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results