accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3NOCl

CAS RN 1421263-60-6

IUPAC Name: N-benzyl-N-tert-butyl-4-chlorobutanamide
Synonyms: N-benzyl-N-tert-butyl-4-chlorobutanamide
Molecular Weight (g/mol): 267.8
Molecular Formula: C15H22ClNO
InChi Key: UBCAUNFPEJQKIA-UHFFFAOYSA-N
SMILES: CC(C)(C)N(CC1=CC=CC=C1)C(=O)CCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results