accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNN

CAS RN 1421263-65-1

IUPAC Name: 7,12-diazatricyclo[6.4.0.0²,⁶]dodeca-1(12),2(6),8,10-tetraen-3-one
Synonyms: 7,12-diazatricyclo[6.4.0.0²,⁶]dodeca-1(12),2(6),8,10-tetraen-3-one
Molecular Weight (g/mol): 172.19
Molecular Formula: C10H8N2O
InChi Key: AMETXLSIVXCORJ-UHFFFAOYSA-N
SMILES: O=C1CCC2=C1C1=NC=CC=C1N2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results