accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3SOONOO

CAS RN 1421263-66-2

IUPAC Name: N-ethyl-N-[(4-methylphenyl)methyl]-4-nitrobenzene-1-sulfonamide
Synonyms: N-ethyl-N-[(4-methylphenyl)methyl]-4-nitrobenzenesulfonamide
Molecular Weight (g/mol): 334.39
Molecular Formula: C16H18N2O4S
InChi Key: IZPJZNVDOFYZRH-UHFFFAOYSA-N
SMILES: CCN(CC1=CC=C(C)C=C1)S(=O)(=O)C1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results