accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1421266-74-1

IUPAC Name: N'-hydroxypiperidine-4-carboximidamide
Synonyms: N'-hydroxypiperidine-4-carboximidamide
Molecular Weight (g/mol): 143.19
Molecular Formula: C6H13N3O
InChi Key: SQZQSQVPENGEOK-UHFFFAOYSA-N
SMILES: NC(=NO)C1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results