accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1422-53-3

IUPAC Name: 2-bromo-4-fluoro-1-methylbenzene
Synonyms: 2-bromo-4-fluoro-1-methylbenzene
Molecular Weight (g/mol): 189.03
Molecular Formula: C7H6BrF
InChi Key: SFGFOJPGCOYQJK-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C=C1Br
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results