accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14220-64-5

IUPAC Name: dichlorobis(phenylformonitrile)palladiumbis(ylium)
Synonyms: dichlorobis(phenylformonitrile)palladiumbis(ylium)
Molecular Weight (g/mol): 383.57
Molecular Formula: C14H10Cl2N2Pd
InChi Key: QQPSWGUJSPTOSH-UHFFFAOYSA-L
SMILES: Cl[Pd++](Cl)([N]#CC1=CC=CC=C1)[N]#CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (4)
  • (2)
  • (1)
Physical Form 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (4)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)