CAS RN 14227-18-0
IUPAC Name:
1,3,5-trimethoxy-2-nitrobenzene
Synonyms:
1,3,5-trimethoxy-2-nitrobenzene
Molecular Weight (g/mol):
213.19
Molecular Formula:
C9H11NO5
InChi Key:
VWYAWLZEMLQGJH-UHFFFAOYSA-N
SMILES:
COC1=CC(OC)=C(C(OC)=C1)[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
1,3,5-Trimethoxy-2-nitrobenzene, 98%
1,3,5-Trimethoxy-2-nitrobenzene, 98%
CAS: 14227-18-0 Molecular Formula: C9H11NO5 Molecular Weight (g/mol): 213.189 MDL Number: MFCD00016992 InChI Key: VWYAWLZEMLQGJH-UHFFFAOYSA-N PubChem CID: 518903 IUPAC Name: 1,3,5-trimethoxy-2-nitrobenzene SMILES: COC1=CC(=C(C(=C1)OC)[N+](=O)[O-])OC