accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1423-26-3

IUPAC Name: [3-(trifluoromethyl)phenyl]boronic acid
Synonyms: 3-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol): 189.93
Molecular Formula: C7H6BF3O2
InChi Key: WOAORAPRPVIATR-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC(=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (6)
  • (1)
Quantity 
  • (3)
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (1)
  • (6)
  • (1)
  • (3)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)