accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrS

CAS RN 1423-61-6

IUPAC Name: 7-bromo-1-benzothiophene
Synonyms: 7-bromo-1-benzothiophene
Molecular Weight (g/mol): 213.09
Molecular Formula: C8H5BrS
InChi Key: NOICDPBEDNMHQK-UHFFFAOYSA-N
SMILES: BrC1=C2SC=CC2=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results