accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14237-71-9

IUPAC Name: 2-chloro-4,6-dimethylpyridine-3-carbonitrile
Synonyms: 2-chloro-4,6-dimethylpyridine-3-carbonitrile
Molecular Weight (g/mol): 166.61
Molecular Formula: C8H7ClN2
InChi Key: RETJKTAVEQPNMH-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C#N)C(Cl)=N1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)