accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1424-66-4

IUPAC Name: 2-chloro-4-(dimethylamino)benzaldehyde
Synonyms: 2-chloro-4-(dimethylamino)benzaldehyde
Molecular Weight (g/mol): 183.64
Molecular Formula: C9H10ClNO
InChi Key: XSQFAWMDRFSIMY-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=O)C(Cl)=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)