accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14243-65-3

IUPAC Name: bromogold; triphenylphosphane
Synonyms: gold(I) bromide; triphenylphosphine
Molecular Weight (g/mol): 539.16
Molecular Formula: C18H15AuBrP
InChi Key: UIYUJCNGYSSFGP-UHFFFAOYSA-M
SMILES: Br[Au].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results