accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SNH2NNH2HBrHBr

CAS RN 142457-00-9

IUPAC Name: 5-(2-aminoethyl)-4-methyl-1,3-thiazol-2-amine dihydrobromide
Synonyms: amthamine dihydrobromide
Molecular Weight (g/mol): 319.06
Molecular Formula: C6H13Br2N3S
InChi Key: XFXNNOPUDSFVJE-UHFFFAOYSA-N
SMILES: Br.Br.CC1=C(CCN)SC(N)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results