CAS RN 14250-96-5
IUPAC Name:
(2E)-2-methylpent-2-enal
Synonyms:
2-pentenal, 2-methyl-
Molecular Weight (g/mol):
98.15
Molecular Formula:
C6H10O
InChi Key:
IDEYZABHVQLHAF-GQCTYLIASA-N
SMILES:
CC\C=C(/C)C=O
1
–
1
of
1
results
1
trans-2-Methyl-2-pentenal 97.0+%, TCI America™
CAS: 14250-96-5 Molecular Formula: C6H10O Molecular Weight (g/mol): 98.145 MDL Number: MFCD00006978 InChI Key: IDEYZABHVQLHAF-GQCTYLIASA-N PubChem CID: 5319754 IUPAC Name: (E)-2-methylpent-2-enal SMILES: CCC=C(C)C=O