accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 142547-15-7

IUPAC Name: ethyl 2-aminocyclopentane-1-carboxylate hydrochloride
Synonyms: ethyl 2-aminocyclopentane-1-carboxylate hydrochloride
Molecular Weight (g/mol): 193.67
Molecular Formula: C8H16ClNO2
InChi Key: RYBMXCZWUCHLJB-UHFFFAOYNA-N
SMILES: Cl.CCOC(=O)C1CCCC1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results