accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 142617-53-6

IUPAC Name: 5-({bis[(2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl)oxy]bismuthanyl}oxy)-2,2,6,6-tetramethylhept-4-en-3-one
Synonyms: 5-({bis[(2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl)oxy]bismuthanyl}oxy)-2,2,6,6-tetramethylhept-4-en-3-one
Molecular Weight (g/mol): 758.79
Molecular Formula: C33H57BiO6
InChi Key: ZNHBPQSSVCRFST-UHFFFAOYSA-K
SMILES: CC(C)(C)C(=O)C=C(O[Bi](OC(=CC(=O)C(C)(C)C)C(C)(C)C)OC(=CC(=O)C(C)(C)C)C(C)(C)C)C(C)(C)C
Boiling Point 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results