accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14267-17-5

NiH3CNH3CSSH3CNCH3SS

CAS RN 14267-17-5

IUPAC Name: nickel(2+) bis((diethylcarbamothioyl)sulfanide)
Synonyms: nickel(2+) didiethyldithiocarbamate
Molecular Weight (g/mol): 355.22
Molecular Formula: C10H20N2NiS4
InChi Key: NCLUCMXMAPDFGT-UHFFFAOYSA-L
SMILES: [Ni++].CCN(CC)C([S-])=S.CCN(CC)C([S-])=S
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)