accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1427-07-2

IUPAC Name: 2-fluoro-1-methyl-4-nitrobenzene
Synonyms: 2-fluoro-1-methyl-4-nitrobenzene
Molecular Weight (g/mol): 155.13
Molecular Formula: C7H6FNO2
InChi Key: WIQISTBTOQNVCE-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1F)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)