accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14271-86-4

IUPAC Name: naphthalene-1-carbonyl cyanide
Synonyms: naphthalene-1-carbonyl cyanide
Molecular Weight (g/mol): 181.19
Molecular Formula: C12H7NO
InChi Key: RITADAJGEQPFIR-UHFFFAOYSA-N
SMILES: O=C(C#N)C1=CC=CC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results