accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOI

CAS RN 14273-85-9

IUPAC Name: methyl 4-iodobutanoate
Synonyms: methyl 4-iodobutanoate
Molecular Weight (g/mol): 228.03
Molecular Formula: C5H9IO2
InChi Key: NBCIIVXSBPDKOM-UHFFFAOYSA-N
SMILES: COC(=O)CCCI
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)