accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 142808-15-9

IUPAC Name: 4-bromo-2-fluoro-1-(trifluoromethyl)benzene
Synonyms: 4-bromo-2-fluoro-1-(trifluoromethyl)benzene
Molecular Weight (g/mol): 243
Molecular Formula: C7H3BrF4
InChi Key: OEPBVXQEVBURGC-UHFFFAOYSA-N
SMILES: FC1=C(C=CC(Br)=C1)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results