accessibility menu, dialog, popup

UserName

Viewing profile as user:
ZnH2NOOH2NOO

CAS RN 14281-83-5

IUPAC Name: zinc(2+) bis(2-aminoacetate)
Synonyms: zinc glycinate
Molecular Weight (g/mol): 213.5
Molecular Formula: C4H8N2O4Zn
InChi Key: UOXSXMSTSYWNMH-UHFFFAOYSA-L
SMILES: [Zn++].NCC([O-])=O.NCC([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results